Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BZIP9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_122389.4 | NP_568457.1 | 3 - 4 | 437 | 91 | |
GenBank mRNA | AF310223.1 | - | 3 - 4 | 433 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |