Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLDALPHA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001343920.1 | NP_001318645.1 | 3 - 4 | 1371 | 86 | |
NM_001343921.1 | NP_001331647.1 | 3 - 4 | 1428 | 86 | ||
NM_001343922.1 | NP_001331648.1 | 1 - 2 | 1288 | 86 | ||
NM_122446.2 | NP_197919.1 | 2 - 3 | 1541 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |