Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ASD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_122513.4 | NP_197984.2 | 11 - 12 | 1262 | 117 | |
GenBank mRNA | AY063925.1 | - | 11 - 12 | 1262 | 117 | |
AY243510.1 | - | 10 - 11 | 1192 | 117 | ||
BT006062.1 | - | 10 - 11 | 1187 | 117 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |