Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLC-D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001343969.1 | NP_001330038.1 | 7 - 8 | 1109 | 82 | |
NM_122525.3 | NP_197996.1 | 7 - 8 | 1124 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |