Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IREG3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_122564.5 | NP_198034.2 | 3 - 4 | 742 | 68 | |
GenBank mRNA | AY062606.1 | - | 3 - 4 | 694 | 68 | |
AY128787.1 | - | 3 - 4 | 453 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |