Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LON1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_122568.4 | NP_568490.4 | 8 - 9 | 1173 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |