Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TPR3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203480.1 | NP_001190409.1 | 4 - 5 | 679 | 66 | |
NM_122585.5 | NP_198055.3 | 4 - 5 | 837 | 66 | ||
GenBank mRNA | BT004037.1 | - | 4 - 5 | 668 | 66 | |
BT005165.1 | - | 4 - 5 | 535 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |