Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SULTR1;3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001332538.1 | NP_001319061.1 | 6 - 7 | 1027 | 74 | |
NM_001332539.1 | NP_001321054.1 | 6 - 7 | 901 | 74 | ||
NM_001332540.1 | NP_001321055.1 | 6 - 7 | 901 | 74 | ||
NM_102065.2 | NP_564159.1 | 5 - 6 | 826 | 74 | ||
GenBank mRNA | AB049624.1 | - | 5 - 6 | 826 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |