Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COX6B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_102094.4 | NP_173661.1 | 4 - 5 | 846 | 104 | |
GenBank mRNA | AB035444.1 | - | 4 - 5 | 548 | 104 | |
AY063997.1 | - | 4 - 5 | 653 | 104 | ||
AY096696.1 | - | 4 - 5 | 520 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |