Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATSAC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_102109.4 | NP_173676.2 | 7 - 8 | 1287 | 76 | |
GenBank mRNA | AF266460.1 | - | 7 - 8 | 1085 | 76 | |
AY227244.1 | - | 7 - 8 | 1085 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |