Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GI | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_102124.3 | NP_564180.1 | 5 - 6 | 1073 | 82 | |
GenBank mRNA | AF105064.1 | - | 4 - 5 | 845 | 82 | |
AJ133786.1 | - | 4 - 5 | 635 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |