Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CEL5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036005.2 | NP_001031082.1 | 3 - 4 | 1364 | 105 | |
NM_102134.3 | NP_173701.1 | 4 - 5 | 1283 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |