Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BCA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001332620.1 | NP_001321762.1 | 4 - 5 | 858 | 78 | |
NM_102221.5 | NP_173785.1 | 5 - 6 | 496 | 78 | ||
GenBank mRNA | AY081265.1 | - | 5 - 6 | 429 | 78 | |
AY114551.1 | - | 4 - 5 | 359 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |