Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_102258.6 | NP_173822.2 | 3 - 4 | 664 | 88 | |
GenBank mRNA | AY074564.1 | - | 3 - 4 | 591 | 88 | |
AY226825.1 | - | 3 - 4 | 596 | 88 | ||
BT025997.1 | - | 3 - 4 | 348 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |