Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALB4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_102294.7 | NP_173858.5 | 2 - 3 | 613 | 93 | |
GenBank mRNA | AM177312.1 | - | 2 - 3 | 574 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |