Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALB4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001160892.1 | NP_001154364.1 | 2 - 3 | 529 | 139 | |
NM_102294.7 | NP_173858.5 | 3 - 4 | 679 | 139 | ||
GenBank mRNA | AM177312.1 | - | 3 - 4 | 640 | 139 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |