Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: THO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001198157.1 | NP_001185086.1 | 17 - 18 | 1877 | 71 | |
NM_001332655.1 | NP_001321786.1 | 16 - 17 | 2068 | 71 | ||
NM_001332656.1 | NP_001321785.1 | 16 - 17 | 2068 | 71 | ||
NM_001332657.1 | NP_001321784.1 | 17 - 18 | 1981 | 71 | ||
NM_001332658.1 | NP_001321783.1 | 17 - 18 | 1818 | 71 | ||
NM_001332659.1 | NP_001321782.1 | 16 - 17 | 2028 | 71 | ||
NM_001332660.1 | NP_001321781.1 | 15 - 16 | 2103 | 71 | ||
NM_001332661.1 | NP_001321780.1 | 11 - 12 | 1400 | 71 | ||
NM_102309.6 | NP_173871.6 | 17 - 18 | 1877 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |