Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OVA9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001198165.1 | NP_001185094.1 | 3 - 4 | 314 | 93 | |
NM_102345.4 | NP_173906.2 | 3 - 4 | 359 | 93 | ||
GenBank mRNA | AY062594.1 | - | 3 - 4 | 313 | 93 | |
AY114653.1 | - | 3 - 4 | 258 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |