Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RCN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_102360.4 | NP_173920.1 | 10 - 11 | 1617 | 62 | |
GenBank mRNA | AY120757.1 | - | 10 - 11 | 1559 | 62 | |
BT000108.1 | - | 9 - 10 | 1442 | 62 | ||
U21557.1 | - | 10 - 11 | 1520 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |