Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GRF12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_102411.3 | NP_564249.1 | 5 - 6 | 986 | 73 | |
GenBank mRNA | AF335544.1 | - | 5 - 6 | 738 | 73 | |
AK117907.1 | - | 5 - 6 | 842 | 73 | ||
BT006230.1 | - | 5 - 6 | 738 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |