Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BGLU40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_102418.4 | NP_173978.1 | 2 - 3 | 586 | 84 | |
GenBank mRNA | AY045927.1 | - | 2 - 3 | 420 | 84 | |
AY142610.1 | - | 2 - 3 | 285 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |