Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MLO14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001332726.1 | NP_001319086.1 | 7 - 8 | 1074 | 84 | |
NM_102433.1 | NP_564257.1 | 6 - 7 | 765 | 84 | ||
GenBank mRNA | AF369575.1 | - | 6 - 7 | 765 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |