Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_102446.3 | NP_564264.1 | 1 - 2 | 229 | 95 | |
GenBank mRNA | AY063844.1 | - | 1 - 2 | 103 | 95 | |
AY088821.1 | - | 1 - 2 | 151 | 95 | ||
AY117288.1 | - | 1 - 2 | 89 | 95 | ||
U05207.1 | - | 1 - 2 | 131 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |