Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PUB45 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_102556.5 | NP_174112.1 | 4 - 5 | 775 | 83 | |
GenBank mRNA | AY094469.1 | - | 4 - 5 | 687 | 83 | |
BT002721.1 | - | 3 - 4 | 343 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |