Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DPL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_102563.4 | NP_174119.1 | 3 - 4 | 716 | 109 | |
GenBank mRNA | AB175035.1 | - | 3 - 4 | 410 | 109 | |
AF360166.1 | - | 3 - 4 | 469 | 109 | ||
AY113914.1 | - | 3 - 4 | 410 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |