Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LEC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_102595.2 | NP_564304.1 | 5 - 6 | 830 | 120 | |
GenBank mRNA | DQ446296.1 | - | 5 - 6 | 792 | 120 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |