Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RLP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_102600.2 | NP_174156.1 | 8 - 9 | 1967 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |