Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ERD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001332846.1 | NP_001321392.1 | 3 - 4 | 626 | 121 | |
NM_102673.6 | NP_564326.1 | 3 - 4 | 796 | 121 | ||
GenBank mRNA | AY093240.1 | - | 3 - 4 | 604 | 121 | |
BT009655.1 | - | 3 - 4 | 470 | 121 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |