Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ESP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001334607.1 | NP_001320490.1 | 1 - 2 | 142 | 88 | |
NM_106044.6 | NP_177525.2 | 1 - 2 | 142 | 88 | ||
GenBank mRNA | AY070424.1 | - | 1 - 2 | 134 | 88 | |
AY096602.1 | - | 1 - 2 | 98 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |