Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ESP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001334607.1 | NP_001320490.1 | 4 - 5 | 962 | 70 | |
NM_106044.6 | NP_177525.2 | 4 - 5 | 818 | 70 | ||
GenBank mRNA | AY070424.1 | - | 4 - 5 | 810 | 70 | |
AY096602.1 | - | 4 - 5 | 774 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |