Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPPA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001334622.1 | NP_001322608.1 | 3 - 4 | 823 | 93 | |
NM_106058.3 | NP_565077.2 | 3 - 4 | 841 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |