Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SOT18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_106069.3 | NP_177549.1 | 1 | 1225 | 90 | |
GenBank mRNA | AK117463.1 | - | 1 | 1094 | 90 | |
BT004984.1 | - | 1 | 1036 | 90 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Both primers and probe map within a single exon |