Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GR-RBP5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_106083.5 | NP_177563.1 | 4 - 5 | 465 | 88 | |
GenBank mRNA | BT002037.1 | - | 4 - 5 | 361 | 88 | |
BT006315.1 | - | 3 - 4 | 292 | 88 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |