Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SFP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_122617.2 | NP_568493.1 | 13 - 14 | 1453 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |