Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SFP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_122617.2 | NP_568493.1 | 17 - 18 | 1734 | 72 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |