Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SFP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001344005.1 | NP_001330511.1 | 3 - 4 | 484 | 74 | |
NM_001344006.1 | NP_001330508.1 | 3 - 4 | 433 | 74 | ||
NM_122618.3 | NP_568494.1 | 3 - 4 | 434 | 74 | ||
GenBank mRNA | AY026255.1 | - | 3 - 4 | 331 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |