Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MIRO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036876.2 | NP_001031953.1 | 4 - 5 | 719 | 69 | |
NM_122636.4 | NP_198106.1 | 4 - 5 | 678 | 69 | ||
GenBank mRNA | AY081280.1 | - | 4 - 5 | 670 | 69 | |
BT003352.1 | - | 3 - 4 | 506 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |