Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LACS7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_122642.5 | NP_198112.2 | 5 - 6 | 547 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |