Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALY1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_122643.3 | NP_198113.2 | 14 - 15 | 2494 | 123 | |
GenBank mRNA | AB493764.1 | - | 13 - 14 | 2263 | 123 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |