Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WRKY74 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_122748.3 | NP_198217.1 | 2 - 3 | 1098 | 73 | |
GenBank mRNA | AF442398.1 | - | 2 - 3 | 839 | 73 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |