Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EGY1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_122913.4 | NP_198372.1 | 1 - 2 | 385 | 100 | |
GenBank mRNA | AY050809.1 | - | 1 - 2 | 338 | 100 | |
BT001978.1 | - | 1 - 2 | 337 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |