Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FIM5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_122961.4 | NP_198420.1 | 5 - 6 | 890 | 141 | |
GenBank mRNA | AY045780.1 | - | 5 - 6 | 868 | 141 | |
AY142570.1 | - | 4 - 5 | 665 | 141 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |