Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_122966.3 | NP_568532.1 | 6 - 7 | 2047 | 66 | |
GenBank mRNA | AB046869.1 | - | 6 - 7 | 1579 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |