Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: YAK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036893.2 | NP_001031970.1 | 13 - 14 | 2012 | 66 | |
NM_001344125.1 | NP_001332507.1 | 13 - 14 | 1490 | 66 | ||
NM_122989.4 | NP_198447.2 | 13 - 14 | 2036 | 66 | ||
GenBank mRNA | AK230350.1 | - | 13 - 14 | 1954 | 66 | |
AY093090.1 | - | 13 - 14 | 2012 | 66 | ||
BT010549.1 | - | 13 - 14 | 1445 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |