Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AHP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001344287.1 | NP_001318703.1 | 1 - 2 | 160 | 108 | |
NM_001344288.1 | NP_001330682.1 | 1 - 2 | 273 | 108 | ||
NM_001344289.1 | NP_001330681.1 | 1 - 2 | 264 | 108 | ||
NM_123296.4 | NP_198750.2 | 1 - 2 | 171 | 108 | ||
GenBank mRNA | AB015143.1 | - | 1 - 2 | 77 | 108 | |
AY050944.1 | - | 1 - 2 | 111 | 108 | ||
AY091174.1 | - | 1 - 2 | 63 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |