Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYB23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_123397.3 | NP_198849.1 | 1 - 2 | 260 | 94 | |
GenBank mRNA | AY519631.1 | - | 1 - 2 | 136 | 94 | |
BT025285.1 | - | 1 - 2 | 136 | 94 | ||
Z68158.1 | - | 1 - 2 | 256 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |