Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DEG13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001344343.1 | NP_001318714.1 | 3 - 4 | 459 | 127 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |