Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001344356.1 | NP_001332643.1 | 8 - 9 | 1695 | 127 | |
NM_001344357.1 | NP_001332642.1 | 8 - 9 | 1695 | 127 | ||
NM_123447.3 | NP_198898.2 | 8 - 9 | 1695 | 127 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |