Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLC-A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036913.2 | NP_001031990.1 | 1 - 2 | 573 | 85 | |
NM_123454.3 | NP_198905.1 | 2 - 3 | 803 | 85 | ||
GenBank mRNA | AF044313.1 | - | 2 - 3 | 624 | 85 | |
AY150506.1 | - | 2 - 3 | 378 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |