Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TOM20-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001344374.1 | NP_001330863.1 | 2 - 3 | 234 | 62 | |
NM_123458.5 | NP_198909.1 | 2 - 3 | 234 | 62 | ||
GenBank mRNA | AF462861.1 | - | 2 - 3 | 231 | 62 | |
AK117822.1 | - | 2 - 3 | 219 | 62 | ||
AY141998.1 | - | 2 - 3 | 168 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |